Product Details

SNP ID
rs111824150
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:104033286 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAGTCACGTTGCTGTAGGCAGAGA[A/G]GACCTTGCTGATGCCGGGTGGCCCC
Phenotype
MIM: 113811 MIM: 616563
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
COL17A1 PubMed Links

Gene Details

Gene
COL17A1
Gene Name
collagen type XVII alpha 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000494.3 4415 Missense Mutation CTC,TTC L1416F NP_000485.3
Gene
SLK
Gene Name
STE20 like kinase
There are no transcripts associated with this gene.

View Full Product Details