Product Details

SNP ID
rs112332349
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:84181781 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTGACTCTATTGGCTCTTTACGGG[C/T]GTCAAGCACTATCCCCAAGCCCAGG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C10orf99 PubMed Links

Gene Details

Gene
C10orf99
Gene Name
chromosome 10 open reading frame 99
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_207373.2 Intron NP_997256.1
XM_005269811.3 Intron XP_005269868.1
Gene
HOST2
Gene Name
human ovarian cancer-specific transcript 2
There are no transcripts associated with this gene.

View Full Product Details