Product Details

SNP ID
rs115611449
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:70205936 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTTGCTAGAACCAAGTCAAGTAAA[C/T]TCCCATTGATTCTTTTGCGGAAATG
Phenotype
MIM: 179030
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PPA1 PubMed Links

Gene Details

Gene
PPA1
Gene Name
pyrophosphatase (inorganic) 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021129.3 Intron NP_066952.1

View Full Product Details