Product Details

SNP ID
rs115923383
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:70301374 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGGCCTAAGGTAGGGGGGCTCTTG[C/T]GCCTTCCGGAGACATGAGCATGTCA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LRRC20 PubMed Links

Gene Details

Gene
LRRC20
Gene Name
leucine rich repeat containing 20
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278211.1 695 Missense Mutation ACA,GCA T179A NP_001265140.1
NM_001278212.1 695 Missense Mutation ACA,GCA T179A NP_001265141.1
NM_001278213.1 695 Missense Mutation ACA,GCA T129A NP_001265142.1
NM_001278214.1 695 Missense Mutation ACA,GCA T110A NP_001265143.1
NM_018205.3 695 Missense Mutation ACA,GCA T123A NP_060675.1
NM_018239.3 695 Missense Mutation ACA,GCA T129A NP_060709.2
NM_207119.2 695 Missense Mutation ACA,GCA T179A NP_997002.1
XM_017016374.1 695 Missense Mutation ACA,GCA T179A XP_016871863.1
XM_017016375.1 695 Missense Mutation ACA,GCA T179A XP_016871864.1

View Full Product Details