Product Details
- SNP ID
-
rs117004549
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:23439901 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTGCCCCGCGCGGCCGCTGCCTCCT[C/G]CTCGCCCCGGCGCCCGCAGCCCCGG
- Phenotype
-
MIM: 612022
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
OTUD1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs566770358] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- OTUD1
- Gene Name
- OTU deubiquitinase 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001145373.2 |
633 |
Silent Mutation |
CTC,CTG |
L148L |
NP_001138845.1 |
View Full Product Details