Product Details

SNP ID
rs117133334
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:114175739 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCAAAAGACTCAGAAGGCAGTTCA[A/G]AGATGCCCCAACTTCTCAAACAGGA
Phenotype
MIM: 605796
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CCDC186 PubMed Links

Gene Details

Gene
CCDC186
Gene Name
coiled-coil domain containing 186
There are no transcripts associated with this gene.

Gene
MIR2110
Gene Name
microRNA 2110
There are no transcripts associated with this gene.

Gene
TDRD1
Gene Name
tudor domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_198795.1 Intron NP_942090.1
XM_005269978.2 Intron XP_005270035.1
XM_005269979.2 Intron XP_005270036.1
XM_011539959.2 Intron XP_011538261.1
XM_011539960.2 Intron XP_011538262.1
XM_011539961.2 Intron XP_011538263.1
XM_011539962.1 Intron XP_011538264.1
XM_011539963.1 Intron XP_011538265.1
XM_011539964.1 Intron XP_011538266.1
XM_011539966.1 Intron XP_011538268.1
XM_017016414.1 Intron XP_016871903.1
XM_017016415.1 Intron XP_016871904.1

View Full Product Details