Product Details

SNP ID
rs118037730
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:103315214 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTTTTTATTCCCCCCAGACAGGGT[A/G]TCACTCTGTCACTCAGGCTGGAGTG
Phenotype
MIM: 607816
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PCGF6 PubMed Links

Gene Details

Gene
PCGF6
Gene Name
polycomb group ring finger 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001011663.1 Intron NP_001011663.1
NM_032154.3 Intron NP_115530.2

View Full Product Details