Product Details

SNP ID
rs137937533
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:73772356 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACATGGCGGCCGGCCCCATTAGGG[C/T]GGTGTTGGTCCTTCTAGGGGTGCTC
Phenotype
MIM: 608842 MIM: 616932 MIM: 607185
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CHCHD1 PubMed Links

Gene Details

Gene
CHCHD1
Gene Name
coiled-coil-helix-coiled-coil-helix domain containing 1
There are no transcripts associated with this gene.

Gene
FUT11
Gene Name
fucosyltransferase 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001284194.1 182 Missense Mutation GCG,GTG A8V NP_001271123.1
NM_173540.2 182 Missense Mutation GCG,GTG A8V NP_775811.2
XM_006717656.3 182 Missense Mutation GCG,GTG A8V XP_006717719.1
Gene
SEC24C
Gene Name
SEC24 homolog C, COPII coat complex component
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004922.3 182 Intron NP_004913.2
NM_198597.2 182 Intron NP_940999.1
XM_011540380.2 182 Intron XP_011538682.1
XM_011540381.2 182 Intron XP_011538683.1
XM_017016967.1 182 Intron XP_016872456.1
XM_017016968.1 182 Intron XP_016872457.1

View Full Product Details