Product Details
- SNP ID
-
rs138633990
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:73256416 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGCTCAGCTTCTAATTGCCGTCCAA[C/T]CTGAAAAAAGTGCAGCAGTTGGTGA
- Phenotype
-
MIM: 611206
MIM: 609204
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CFAP70
PubMed Links
Gene Details
- Gene
- CFAP70
- Gene Name
- cilia and flagella associated protein 70
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_145170.3 |
3322 |
Missense Mutation |
ATT,GTT |
I1080V |
NP_660153.3 |
XM_005269489.2 |
3322 |
Missense Mutation |
ATT,GTT |
I888V |
XP_005269546.1 |
XM_006717604.2 |
3322 |
Missense Mutation |
ATT,GTT |
I1080V |
XP_006717667.1 |
XM_006717605.2 |
3322 |
Missense Mutation |
ATT,GTT |
I1080V |
XP_006717668.1 |
XM_006717609.2 |
3322 |
Missense Mutation |
ATT,GTT |
I888V |
XP_006717672.1 |
XM_006717610.2 |
3322 |
Missense Mutation |
ATT,GTT |
I888V |
XP_006717673.1 |
XM_006717611.3 |
3322 |
Missense Mutation |
ATT,GTT |
I888V |
XP_006717674.1 |
XM_011539212.2 |
3322 |
Missense Mutation |
ATT,GTT |
I888V |
XP_011537514.1 |
XM_017015620.1 |
3322 |
Missense Mutation |
ATT,GTT |
I1102V |
XP_016871109.1 |
XM_017015621.1 |
3322 |
Missense Mutation |
ATT,GTT |
I1101V |
XP_016871110.1 |
XM_017015622.1 |
3322 |
Missense Mutation |
ATT,GTT |
I1101V |
XP_016871111.1 |
XM_017015623.1 |
3322 |
Missense Mutation |
ATT,GTT |
I1076V |
XP_016871112.1 |
XM_017015624.1 |
3322 |
Missense Mutation |
ATT,GTT |
I1075V |
XP_016871113.1 |
XM_017015625.1 |
3322 |
Missense Mutation |
ATT,GTT |
I1072V |
XP_016871114.1 |
XM_017015626.1 |
3322 |
Missense Mutation |
ATT,GTT |
I1038V |
XP_016871115.1 |
XM_017015627.1 |
3322 |
Missense Mutation |
ATT,GTT |
I1032V |
XP_016871116.1 |
XM_017015628.1 |
3322 |
Missense Mutation |
ATT,GTT |
I1010V |
XP_016871117.1 |
XM_017015629.1 |
3322 |
Missense Mutation |
ATT,GTT |
I967V |
XP_016871118.1 |
XM_017015630.1 |
3322 |
Missense Mutation |
ATT,GTT |
I888V |
XP_016871119.1 |
XM_017015631.1 |
3322 |
Missense Mutation |
ATT,GTT |
I888V |
XP_016871120.1 |
XM_017015632.1 |
3322 |
Missense Mutation |
ATT,GTT |
I887V |
XP_016871121.1 |
XM_017015633.1 |
3322 |
Intron |
|
|
XP_016871122.1 |
- Gene
- DNAJC9
- Gene Name
- DnaJ heat shock protein family (Hsp40) member C9
There are no transcripts associated with this gene.
- Gene
- DNAJC9-AS1
- Gene Name
- DNAJC9 antisense RNA 1
There are no transcripts associated with this gene.
- Gene
- MRPS16
- Gene Name
- mitochondrial ribosomal protein S16
There are no transcripts associated with this gene.
View Full Product Details