Product Details

SNP ID
rs139504082
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:23441037 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTACTGTAGGTGTAATGCCTTAAT[C/T]ATCTTTTTGAATGTTTTCTCAGAGC
Phenotype
MIM: 612022
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OTUD1 PubMed Links

Gene Details

Gene
OTUD1
Gene Name
OTU deubiquitinase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145373.2 1769 UTR 3 NP_001138845.1

View Full Product Details