Product Details
- SNP ID
-
rs140318221
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:121479866 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGGCAGACACAGTCATTCATGTTTT[A/G]ACACTGCCGTTTATGTGTGGATACT
- Phenotype
-
MIM: 176943
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
FGFR2
PubMed Links
Gene Details
- Gene
- FGFR2
- Gene Name
- fibroblast growth factor receptor 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_000141.4 |
3391 |
Silent Mutation |
GTC,GTT |
V819V |
NP_000132.3 |
NM_001144913.1 |
3391 |
Intron |
|
|
NP_001138385.1 |
NM_001144914.1 |
3391 |
Silent Mutation |
GTC,GTT |
V707V |
NP_001138386.1 |
NM_001144915.1 |
3391 |
Intron |
|
|
NP_001138387.1 |
NM_001144916.1 |
3391 |
Silent Mutation |
GTC,GTT |
V704V |
NP_001138388.1 |
NM_001144917.1 |
3391 |
Silent Mutation |
GTC,GTT |
V703V |
NP_001138389.1 |
NM_001144918.1 |
3391 |
Silent Mutation |
GTC,GTT |
V702V |
NP_001138390.1 |
NM_001144919.1 |
3391 |
Intron |
|
|
NP_001138391.1 |
NM_001320654.1 |
3391 |
Silent Mutation |
GTC,GTT |
V591V |
NP_001307583.1 |
NM_001320658.1 |
3391 |
Silent Mutation |
GTC,GTT |
V817V |
NP_001307587.1 |
NM_022970.3 |
3391 |
Silent Mutation |
GTC,GTT |
V820V |
NP_075259.4 |
NM_023029.2 |
3391 |
Silent Mutation |
GTC,GTT |
V730V |
NP_075418.1 |
XM_006717708.2 |
3391 |
Silent Mutation |
GTC,GTT |
V837V |
XP_006717771.1 |
XM_006717710.3 |
3391 |
UTR 3 |
|
|
XP_006717773.1 |
XM_006717711.2 |
3391 |
Silent Mutation |
GTC,GTT |
V750V |
XP_006717774.1 |
XM_006717712.2 |
3391 |
Silent Mutation |
GTC,GTT |
V724V |
XP_006717775.1 |
XM_017015920.1 |
3391 |
UTR 3 |
|
|
XP_016871409.1 |
XM_017015921.1 |
3391 |
UTR 3 |
|
|
XP_016871410.1 |
XM_017015922.1 |
3391 |
Silent Mutation |
GTC,GTT |
V748V |
XP_016871411.1 |
XM_017015923.1 |
3391 |
Silent Mutation |
GTC,GTT |
V747V |
XP_016871412.1 |
XM_017015924.1 |
3391 |
Silent Mutation |
GTC,GTT |
V723V |
XP_016871413.1 |
XM_017015925.1 |
3391 |
UTR 3 |
|
|
XP_016871414.1 |
View Full Product Details