Product Details
- SNP ID
-
rs141025438
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:124448039 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCGCGCTAGGCCCGGCAGCAAGCGC[A/G]CAGCCCGCTCCCGCAGCCGCGGCCT
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
NKX1-2
PubMed Links
Gene Details
- Gene
- NKX1-2
- Gene Name
- NK1 homeobox 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001146340.1 |
361 |
Missense Mutation |
GCG,GTG |
A108V |
NP_001139812.1 |
View Full Product Details