Product Details

SNP ID
rs141565261
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:68284254 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCCTTCCGTCTGGACGAAGGGAAA[C/T]TCCCAGCTCTCCTCCTCGGTGGGAA
Phenotype
MIM: 612189
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PBLD PubMed Links

Gene Details

Gene
PBLD
Gene Name
phenazine biosynthesis like protein domain containing
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001033083.1 961 Intron NP_001028255.1
NM_022129.3 961 Missense Mutation ATT,GTT I264V NP_071412.2
XM_005270028.3 961 Missense Mutation ATT,GTT I264V XP_005270085.1
XM_011540060.2 961 Silent Mutation GAA,GAG E255E XP_011538362.1
XM_017016513.1 961 Silent Mutation GAA,GAG E255E XP_016872002.1
XM_017016514.1 961 Intron XP_016872003.1

View Full Product Details