Product Details

SNP ID
rs141695177
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.10:32015643 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGGAGAAGCTGCTGGATGTTGCCC[A/G]GGACGAATAGGTTTAGCTAATATGA
Phenotype
MIM: 602809
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
KIF5B PubMed Links

Gene Details

Gene
KIF5B
Gene Name
kinesin family member 5B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004521.2 2854 Silent Mutation CCC,CCT P926P NP_004512.1
XM_017016224.1 2854 Silent Mutation CCC,CCT P832P XP_016871713.1

View Full Product Details