Product Details

SNP ID
rs141904814
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:93062935 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCGCCACGCTGGCCCGGACCTTCGA[A/G]CAGCTCGTGGAGAACCTCTTCTCAC
Phenotype
MIM: 608428 MIM: 609672
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CYP26C1 PubMed Links

Gene Details

Gene
CYP26C1
Gene Name
cytochrome P450 family 26 subfamily C member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_183374.2 645 Silent Mutation GAA,GAG E215E NP_899230.2
Gene
EXOC6
Gene Name
exocyst complex component 6
There are no transcripts associated with this gene.

View Full Product Details