Product Details

SNP ID
rs142094831
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:21756729 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTTCTTTTGGACCAGTTCAACCAG[A/C]AACTTGTACCTAGCGATACAGTCTT
Phenotype
MIM: 611207
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
DNAJC1 PubMed Links

Gene Details

Gene
DNAJC1
Gene Name
DnaJ heat shock protein family (Hsp40) member C1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022365.3 1910 Missense Mutation TTG,TTT L541F NP_071760.2
XM_011519614.2 1910 Intron XP_011517916.1
XM_017016536.1 1910 Intron XP_016872025.1

View Full Product Details