Product Details

SNP ID
rs142205547
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:66927952 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATTATCTGTGCCAGTCCCAAAGAG[C/T]TGCAAGGAGTAAATGTGATCGATGC
Phenotype
MIM: 607667 MIM: 610869
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CTNNA3 PubMed Links

Gene Details

Gene
CTNNA3
Gene Name
catenin alpha 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001127384.2 1586 Intron NP_001120856.1
NM_001291133.1 1586 Intron NP_001278062.1
NM_013266.3 1586 Intron NP_037398.2
XM_017016151.1 1586 Intron XP_016871640.1
XM_017016152.1 1586 Intron XP_016871641.1
XM_017016153.1 1586 Intron XP_016871642.1
XM_017016154.1 1586 Intron XP_016871643.1
XM_017016155.1 1586 Intron XP_016871644.1
XM_017016156.1 1586 Intron XP_016871645.1
XM_017016157.1 1586 Intron XP_016871646.1
XM_017016158.1 1586 Intron XP_016871647.1
Gene
LOC101928961
Gene Name
uncharacterized LOC101928961
There are no transcripts associated with this gene.

Gene
LRRTM3
Gene Name
leucine rich repeat transmembrane neuronal 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_178011.4 1586 Silent Mutation CTG,TTG L346L NP_821079.3

View Full Product Details