Product Details

SNP ID
rs142543885
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:17953503 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGTTGGAGAAAACTGGGTGCCCAC[A/G]GAGGAGAAACGGAATGCAAGGAGAT
Phenotype
MIM: 608734
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SLC39A12 PubMed Links

Gene Details

Gene
SLC39A12
Gene Name
solute carrier family 39 member 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145195.1 500 Missense Mutation CAG,CGG Q76R NP_001138667.1
NM_001282733.1 500 Missense Mutation CAG,CGG Q76R NP_001269662.1
NM_001282734.1 500 Intron NP_001269663.1
NM_152725.3 500 Missense Mutation CAG,CGG Q76R NP_689938.2

View Full Product Details