Product Details

SNP ID
rs142606828
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:70598447 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTGAACCAGTCCCCCCACAGGCCC[C/T]ATGCTTGGATGAAGGTCACCTCCAG
Phenotype
MIM: 170280
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PRF1 PubMed Links

Gene Details

Gene
PRF1
Gene Name
perforin 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001083116.1 1435 Nonsense Mutation TAG,TGG *425W NP_001076585.1
NM_005041.4 1435 Nonsense Mutation TAG,TGG *425W NP_005032.2

View Full Product Details