Product Details

SNP ID
rs142735682
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:59176890 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCGCCTGGATCATGCCCTCAACAGC[C/G]CCACCAGCCCCTGTGAGGAGGTGAT
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CCEPR PubMed Links

Gene Details

Gene
CCEPR
Gene Name
cervical carcinoma expressed PCNA regulatory lncRNA
There are no transcripts associated with this gene.

Gene
PHYHIPL
Gene Name
phytanoyl-CoA 2-hydroxylase interacting protein like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001143774.1 274 Intron NP_001137246.1
NM_032439.3 274 Missense Mutation CCC,GCC P13A NP_115815.2
XM_011540275.2 274 UTR 5 XP_011538577.1
XM_011540276.2 274 Intron XP_011538578.1
XM_017016782.1 274 Intron XP_016872271.1
XM_017016783.1 274 Intron XP_016872272.1

View Full Product Details