Product Details

SNP ID
rs142954568
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:1159964 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGGCAGCACAGGAATTGGCACAAC[C/G]TGAAGTCAGCTCTGCAGCTTTCCAA
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
LINC00200 PubMed Links

Gene Details

Gene
LINC00200
Gene Name
long intergenic non-protein coding RNA 200
There are no transcripts associated with this gene.

View Full Product Details