Product Details

SNP ID
rs144051413
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:133381550 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCCTGTGAAGACCATCCACTGGAA[C/T]GGGTCCTTCCAGGAGGCAGCCTTTC
Phenotype
MIM: 602292 MIM: 615853
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ECHS1 PubMed Links

Gene Details

Gene
ECHS1
Gene Name
enoyl-CoA hydratase, short chain 1
There are no transcripts associated with this gene.

Gene
MIR3944
Gene Name
microRNA 3944
There are no transcripts associated with this gene.

Gene
PAOX
Gene Name
polyamine oxidase (exo-N4-amino)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152911.3 839 Silent Mutation AAC,AAT N253N NP_690875.1
NM_207127.2 839 Silent Mutation AAC,AAT N253N NP_997010.1
NM_207128.2 839 Silent Mutation AAC,AAT N253N NP_997011.1

View Full Product Details