Product Details

SNP ID
rs144913185
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:15078581 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTTTGCTTTAGAAATATAGGCACT[C/T]GTCGCATCTTCCGTCGACAACCCTA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ACBD7 PubMed Links

Gene Details

Gene
ACBD7
Gene Name
acyl-CoA binding domain containing 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039844.2 Intron NP_001034933.1
Gene
OLAH
Gene Name
oleoyl-ACP hydrolase
There are no transcripts associated with this gene.

View Full Product Details