Product Details

SNP ID
hCV160629806
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:47322983 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGTCCGATAAGTCGACTACGCCAGC[G/A]TCCAACATTGTGCGGAGCTTCAGCA
Phenotype
MIM: 601361 MIM: 605120
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
GDF10 PubMed Links

Gene Details

Gene
GDF10
Gene Name
growth differentiation factor 10
There are no transcripts associated with this gene.

Gene
GDF2
Gene Name
growth differentiation factor 2
There are no transcripts associated with this gene.

View Full Product Details