Product Details

SNP ID
rs145226223
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:5499276 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCGCCGTCGCCGTCGCTGTCAACCT[C/T]GGAGATGAGTTTCCTTAGCTGGGCC
Phenotype
MIM: 605183
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CALML5 PubMed Links

Gene Details

Gene
CALML5
Gene Name
calmodulin like 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017422.4 295 Missense Mutation AAG,GAG K55E NP_059118.2
Gene
LOC105376379
Gene Name
uncharacterized LOC105376379
There are no transcripts associated with this gene.

View Full Product Details