Product Details

SNP ID
rs145770636
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:12898650 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTTGACGTGGTCCTCCTCCTCAAA[C/T]AAGGCCTTCTGCACCGAGGAGCACA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CCDC3 PubMed Links

Gene Details

Gene
CCDC3
Gene Name
coiled-coil domain containing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282658.1 713 Silent Mutation TTA,TTG L68L NP_001269587.1
NM_031455.3 713 Silent Mutation TTA,TTG L193L NP_113643.1

View Full Product Details