Product Details

SNP ID
rs146369046
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.10:43154175 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAAAATCTCTCTCTCCAGTAAACC[A/G]TTATTACTGTAACAGAAACAGTTTC
Phenotype
MIM: 616616
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
CSGALNACT2 PubMed Links

Gene Details

Gene
CSGALNACT2
Gene Name
chondroitin sulfate N-acetylgalactosaminyltransferase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001319654.1 Intron NP_001306583.1
NM_001319656.1 Intron NP_001306585.1
NM_018590.4 Intron NP_061060.3
XM_011539938.1 Intron XP_011538240.1
XM_017016388.1 Intron XP_016871877.1
XM_017016389.1 Intron XP_016871878.1
XM_017016390.1 Intron XP_016871879.1
XM_017016391.1 Intron XP_016871880.1

View Full Product Details