Product Details

SNP ID
rs146373216
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:93566807 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCGCACCCGCTTTCCCTTCTTCTC[C/T]GACGTCAAGGGCGACCACCGGCTGG
Phenotype
MIM: 609044
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FFAR4 PubMed Links

Gene Details

Gene
FFAR4
Gene Name
free fatty acid receptor 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001195755.1 629 Silent Mutation TCC,TCT S29S NP_001182684.1
NM_181745.3 629 Silent Mutation TCC,TCT S29S NP_859529.2
XM_011539746.2 629 Silent Mutation TCC,TCT S29S XP_011538048.1

View Full Product Details