Product Details

SNP ID
rs146505608
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:84248063 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTAGTTCTGGGCTTCTTCCATAAAT[C/T]GGATATTATTTCCTAACTCATATAG
Phenotype
MIM: 616103 MIM: 600342
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LRIT1 PubMed Links

Gene Details

Gene
LRIT1
Gene Name
leucine rich repeat, Ig-like and transmembrane domains 1
There are no transcripts associated with this gene.

Gene
RGR
Gene Name
retinal G protein coupled receptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001012720.1 Intron NP_001012738.1
NM_001012722.1 Intron NP_001012740.1
NM_002921.3 Intron NP_002912.2

View Full Product Details