Product Details

SNP ID
rs146658749
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:7705162 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCCCCAGGCAAATTTCAATTGGTG[A/G]CAGAGAACCGGAGATATCAGGTATA
Phenotype
MIM: 146640
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ITIH2 PubMed Links

Gene Details

Gene
ITIH2
Gene Name
inter-alpha-trypsin inhibitor heavy chain 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002216.2 301 Missense Mutation ACA,GCA T47A NP_002207.2

View Full Product Details