Product Details

SNP ID
rs147929533
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:50806785 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCACTGCAAAAGTTGGGTAGACCT[C/T]ATAGGTTGTATATGCTGCTAAGTCT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
A1CF PubMed Links

Gene Details

Gene
A1CF
Gene Name
APOBEC1 complementation factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001198818.1 1986 Missense Mutation NP_001185747.1
NM_001198819.1 1986 Missense Mutation NP_001185748.1
NM_001198820.1 1986 Missense Mutation NP_001185749.1
NM_014576.3 1986 Missense Mutation NP_055391.2
NM_138932.2 1986 Missense Mutation NP_620310.1
NM_138933.2 1986 Missense Mutation NP_620311.1
XM_005269718.2 1986 Nonsense Mutation XP_005269775.1
XM_005269720.3 1986 Nonsense Mutation XP_005269777.1
XM_011539729.2 1986 Nonsense Mutation XP_011538031.1
XM_011539730.2 1986 Nonsense Mutation XP_011538032.1
XM_017016160.1 1986 Nonsense Mutation XP_016871649.1

View Full Product Details