Product Details
- SNP ID
-
rs148145718
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:45461377 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCTGTGCAGTGGCAGGGGGTGATCA[G/T]GGGGCTCTCATCATCTCCTTCACAG
- Phenotype
-
MIM: 613335
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
LOC102724323
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs3764990] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- LOC102724323
- Gene Name
- uncharacterized LOC102724323
There are no transcripts associated with this gene.
- Gene
- MARCH8
- Gene Name
- membrane associated ring-CH-type finger 8
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001002266.2 |
1874 |
Missense Mutation |
ATG,CTG |
M93L |
NP_001002266.1 |
NM_001282866.1 |
1874 |
Missense Mutation |
ATG,CTG |
M375L |
NP_001269795.1 |
NM_145021.5 |
1874 |
Missense Mutation |
ATG,CTG |
M93L |
NP_659458.2 |
XM_005271804.2 |
1874 |
Missense Mutation |
ATG,CTG |
M375L |
XP_005271861.1 |
XM_006717704.2 |
1874 |
Missense Mutation |
ATG,CTG |
M375L |
XP_006717767.1 |
XM_011539492.2 |
1874 |
Missense Mutation |
ATG,CTG |
M375L |
XP_011537794.1 |
XM_011539493.2 |
1874 |
Missense Mutation |
ATG,CTG |
M375L |
XP_011537795.1 |
XM_011539494.2 |
1874 |
Missense Mutation |
ATG,CTG |
M375L |
XP_011537796.1 |
XM_011539495.1 |
1874 |
Missense Mutation |
ATG,CTG |
M93L |
XP_011537797.1 |
XM_017015894.1 |
1874 |
Missense Mutation |
ATG,CTG |
M93L |
XP_016871383.1 |
View Full Product Details