Product Details

SNP ID
rs148481159
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:59176867 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGGTTGGAAAAATGGAGGTGCCGC[A/G]CCTGGATCATGCCCTCAACAGCCCC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CCEPR PubMed Links

Gene Details

Gene
CCEPR
Gene Name
cervical carcinoma expressed PCNA regulatory lncRNA
There are no transcripts associated with this gene.

Gene
PHYHIPL
Gene Name
phytanoyl-CoA 2-hydroxylase interacting protein like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001143774.1 251 Intron NP_001137246.1
NM_032439.3 251 Missense Mutation CAC,CGC H5R NP_115815.2
XM_011540275.2 251 UTR 5 XP_011538577.1
XM_011540276.2 251 Intron XP_011538578.1
XM_017016782.1 251 Intron XP_016872271.1
XM_017016783.1 251 Intron XP_016872272.1

View Full Product Details