Product Details

SNP ID
rs148691180
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:97358963 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTACCTTGGCCTTGTATTCAGCCC[C/T]AGGCATAGCCTTCTTGGCCACAGGG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RRP12 PubMed Links

Gene Details

Gene
RRP12
Gene Name
ribosomal RNA processing 12 homolog
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145114.1 3827 Missense Mutation AGG,GGG R1169G NP_001138586.1
NM_001284337.1 3827 Missense Mutation AGG,GGG R1130G NP_001271266.1
NM_015179.3 3827 Missense Mutation AGG,GGG R1230G NP_055994.2
XM_011539555.1 3827 Missense Mutation AGG,GGG R1202G XP_011537857.1
XM_011539556.2 3827 Intron XP_011537858.1

View Full Product Details