Product Details

SNP ID
rs150721203
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:12897925 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGATTTTCAGGCATCTTGGATGCAA[A/G]GCCACCATCCCCTTCCAAAGATGTG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CCDC3 PubMed Links

Gene Details

Gene
CCDC3
Gene Name
coiled-coil domain containing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282658.1 1438 UTR 3 NP_001269587.1
NM_031455.3 1438 UTR 3 NP_113643.1

View Full Product Details