Product Details

SNP ID
rs150772711
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:49735271 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAGGCCTGGAGATTGAAGGCAGTA[C/T]CCAGAAACTTCTTCAGTGACACCAG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OGDHL PubMed Links

Gene Details

Gene
OGDHL
Gene Name
oxoglutarate dehydrogenase-like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001143996.1 3017 Missense Mutation GAT,GGT D940G NP_001137468.1
NM_001143997.1 3017 Missense Mutation GAT,GGT D788G NP_001137469.1
NM_018245.2 3017 Missense Mutation GAT,GGT D997G NP_060715.2
XM_011539946.1 3017 Missense Mutation GAT,GGT D997G XP_011538248.1
XM_011539947.1 3017 Missense Mutation GAT,GGT D997G XP_011538249.1
XM_017016401.1 3017 Missense Mutation GAT,GGT D788G XP_016871890.1
XM_017016402.1 3017 Missense Mutation GAT,GGT D658G XP_016871891.1
XM_017016403.1 3017 Missense Mutation GAT,GGT D658G XP_016871892.1

View Full Product Details