Product Details

SNP ID
rs150953240
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:15096606 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTGTCCCCGACTTTGCAGCGACTC[A/C]GATGGTAGCCTGAGTCTCCATTGCT
Phenotype
MIM: 603801 MIM: 606116
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ACBD7 PubMed Links

Gene Details

Gene
ACBD7
Gene Name
acyl-CoA binding domain containing 7
There are no transcripts associated with this gene.

Gene
C10orf111
Gene Name
chromosome 10 open reading frame 111
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153244.1 493 Silent Mutation NP_694976.1
Gene
NMT2
Gene Name
N-myristoyltransferase 2
There are no transcripts associated with this gene.

Gene
RPP38
Gene Name
ribonuclease P/MRP subunit p38
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001097590.2 493 Intron NP_001091059.1
NM_001265601.1 493 Intron NP_001252530.1
NM_006414.4 493 Intron NP_006405.2
NM_183005.4 493 Intron NP_892117.1
XM_006717363.1 493 Intron XP_006717426.1
XM_006717364.3 493 Intron XP_006717427.1
XM_011519293.1 493 Intron XP_011517595.1
XM_017015479.1 493 Intron XP_016870968.1
XM_017015480.1 493 Intron XP_016870969.1
XM_017015481.1 493 Intron XP_016870970.1

View Full Product Details