Product Details

SNP ID
rs111230950
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.11:4947048 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGTGCATATTCTAGCCCTGGCATC[C/G]CAACCAAGAAGAAGGTGGTGATTTC
Phenotype
Polymorphism
C/G, Transversion Substitution
Allele Nomenclature
Literature Links
OR51A2 PubMed Links
Additional Information
For this assay, SNP(s) [rs111741316] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
OR51A2
Gene Name
olfactory receptor family 51 subfamily A member 2
There are no transcripts associated with this gene.

Gene
OR51A4
Gene Name
olfactory receptor family 51 subfamily A member 4
There are no transcripts associated with this gene.

View Full Product Details