Product Details

SNP ID
rs111308906
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.11:65172483 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGACCTGGAGGGCCCCAAATGTGAG[A/G]TTTTTCCATGGGCCCTGGGAAAAGA
Phenotype
MIM: 114220 MIM: 614030
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
CAPN1 PubMed Links

Gene Details

Gene
CAPN1
Gene Name
calpain 1
There are no transcripts associated with this gene.

Gene
SPDYC
Gene Name
speedy/RINGO cell cycle regulator family member C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001008778.1 788 Missense Mutation ATT,GTT I132V NP_001008778.1
XM_017017705.1 788 Missense Mutation ATT,GTT I113V XP_016873194.1
XM_017017706.1 788 Missense Mutation ATT,GTT I113V XP_016873195.1
XM_017017707.1 788 Missense Mutation ATT,GTT I113V XP_016873196.1
XM_017017708.1 788 Missense Mutation ATT,GTT I113V XP_016873197.1
XM_017017709.1 788 Missense Mutation ATT,GTT I113V XP_016873198.1
XM_017017710.1 788 Missense Mutation ATT,GTT I113V XP_016873199.1

View Full Product Details