Product Details

SNP ID
rs112765358
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:69773453 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGTTGTAGTTGTTGGGAAGGAGAAT[C/T]TCCTTGAACGTGCACTCATCGGTGA
Phenotype
MIM: 164980
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FGF4 PubMed Links

Gene Details

Gene
FGF4
Gene Name
fibroblast growth factor 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002007.2 796 Silent Mutation GAA,GAG E159E NP_001998.1
XM_005273847.1 796 UTR 3 XP_005273904.1

View Full Product Details