Product Details

SNP ID
rs112949144
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:64988171 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAACTGAACTTCCACCCGGTCATGG[A/G]CCCTGCTCTTCCTGCTGTCAGCCTA
Phenotype
MIM: 614983
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BATF2 PubMed Links

Gene Details

Gene
BATF2
Gene Name
basic leucine zipper ATF-like transcription factor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001300807.1 1914 UTR 3 NP_001287736.1
NM_001300808.1 1914 UTR 3 NP_001287737.1
NM_138456.3 1914 UTR 3 NP_612465.3

View Full Product Details