Product Details

SNP ID
rs114500482
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:65945750 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCCGTCGGTGCGACCAGGGCAGGA[C/G]GTGCGGCTCCAGGCTCCAGGAACCA
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
TSGA10IP PubMed Links

Gene Details

Gene
TSGA10IP
Gene Name
testis specific 10 interacting protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152762.2 306 Silent Mutation GAC,GAG D25E NP_689975.2
XM_011544887.2 306 Missense Mutation GAC,GAG D25E XP_011543189.1
XM_011544888.2 306 UTR 5 XP_011543190.1

View Full Product Details