Product Details

SNP ID
rs114893240
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:60927691 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCACCAACGGCCAGTCCCATGGGA[C/T]GTGCGGGCCGTTTCAGTGGAAGCGG
Phenotype
MIM: 610408
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SLC15A3 PubMed Links

Gene Details

Gene
SLC15A3
Gene Name
solute carrier family 15 member 3
There are no transcripts associated with this gene.

Gene
TMEM109
Gene Name
transmembrane protein 109
There are no transcripts associated with this gene.

Gene
TMEM132A
Gene Name
transmembrane protein 132A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017870.3 559 Silent Mutation GAC,GAT D122D NP_060340.2
NM_178031.2 559 Silent Mutation GAC,GAT D122D NP_821174.1
XM_017017951.1 559 Silent Mutation GAC,GAT D135D XP_016873440.1
XM_017017952.1 559 Silent Mutation GAC,GAT D135D XP_016873441.1

View Full Product Details