Product Details

SNP ID
rs114953123
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:65882205 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGTTTGGCCAGCTCTATGGCTATC[A/G]GAGGGCAGCTGGAGGGGTCCCCAGC
Phenotype
MIM: 605360 MIM: 602364 MIM: 604633 MIM: 608296 MIM: 136515
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CCDC85B PubMed Links

Gene Details

Gene
CCDC85B
Gene Name
coiled-coil domain containing 85B
There are no transcripts associated with this gene.

Gene
CTSW
Gene Name
cathepsin W
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001335.3 359 Missense Mutation CAG,CGG Q106R NP_001326.2
Gene
EFEMP2
Gene Name
EGF containing fibulin like extracellular matrix protein 2
There are no transcripts associated with this gene.

Gene
FIBP
Gene Name
FGF1 intracellular binding protein
There are no transcripts associated with this gene.

Gene
FOSL1
Gene Name
FOS like 1, AP-1 transcription factor subunit
There are no transcripts associated with this gene.

View Full Product Details