Product Details

SNP ID
rs115557428
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:86948600 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGATCAAATACACTTTGCCCTGAAA[A/G]GGCAAGAGATTTGTGGAAAAGGCAT
Phenotype
MIM: 604579
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FZD4 PubMed Links

Gene Details

Gene
FZD4
Gene Name
frizzled class receptor 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012193.3 4469 UTR 3 NP_036325.2
Gene
LOC100506368
Gene Name
uncharacterized LOC100506368
There are no transcripts associated with this gene.

Gene
PRSS23
Gene Name
protease, serine 23
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001293178.1 4469 Intron NP_001280107.1
NM_001293179.1 4469 Intron NP_001280108.1
NM_001293180.1 4469 Intron NP_001280109.1
NM_007173.5 4469 Intron NP_009104.2

View Full Product Details