Product Details
- SNP ID
-
rs116188604
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:62192164 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCCAAATTTAAGGCACCTCTGGAAG[C/T]TGTTGCAGCCAAGATGGAAGTGAAG
- Phenotype
-
MIM: 615060
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
SCGB1D1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs3831484] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- SCGB1D1
- Gene Name
- secretoglobin family 1D member 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_006552.1 |
211 |
Missense Mutation |
GCT,GTT |
A55V |
NP_006543.1 |
View Full Product Details