Product Details

SNP ID
rs117096937
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:67519883 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGCTGAGGCGCTCCCCCAGCAGGG[C/T]CTTGAGGGCCGCCCGGAGCTCGCCC
Phenotype
MIM: 607314
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CABP2 PubMed Links

Gene Details

Gene
CABP2
Gene Name
calcium binding protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318496.1 653 Missense Mutation ACC,GCC T189A NP_001305425.1
NM_016366.2 653 Missense Mutation ACC,GCC T183A NP_057450.2

View Full Product Details