Product Details

SNP ID
rs117654845
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:93784345 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTGCGTTCTGGGAAAGTTGCTGGG[C/T]CAGCTCCTTTGTTTCCAGTCTGAGC
Phenotype
MIM: 603810
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MED17 PubMed Links

Gene Details

Gene
MED17
Gene Name
mediator complex subunit 17
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004268.4 107 UTR 5 NP_004259.3
XM_011543068.2 107 UTR 5 XP_011541370.1

View Full Product Details