Product Details

SNP ID
rs117732556
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:94975727 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTATCCTCTGCATATAGAACAGTG[A/C]CTGGCACATAATAGGTGCTGAGTAA
Phenotype
MIM: 609766
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CWC15 PubMed Links

Gene Details

Gene
CWC15
Gene Name
CWC15 spliceosome-associated protein
There are no transcripts associated with this gene.

Gene
KDM4D
Gene Name
lysine demethylase 4D
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018039.2 462 UTR 5 NP_060509.2

View Full Product Details