Product Details

SNP ID
rs138593335
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:68013562 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGGCTACTTAGCAAAACACCCAGA[A/G]CTGACCTTTGTCGAGTACTTTCTGT
Phenotype
MIM: 600466 MIM: 608204
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ALDH3B1 PubMed Links

Gene Details

Gene
ALDH3B1
Gene Name
aldehyde dehydrogenase 3 family member B1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000694.3 Intron NP_000685.1
NM_001030010.2 Intron NP_001025181.1
NM_001161473.2 Intron NP_001154945.1
NM_001290058.1 Intron NP_001276987.1
NM_001290059.1 Intron NP_001276988.1
Gene
UNC93B1
Gene Name
unc-93 homolog B1 (C. elegans)
There are no transcripts associated with this gene.

View Full Product Details